Merck
Dual nitrogen and oxygen isotope fractionation during anaerobic ammonium oxidation by anammox bacteria.
The ISME journal (2019-05-30)
Kanae Kobayashi, Akiko Makabe, Midori Yano, Mamoru Oshiki, Tomonori Kindaichi, Karen L Casciotti, Satoshi Okabe
RESUMEN
Natural abundance of stable nitrogen (N) and oxygen (O) isotopes are invaluable biogeochemical tracers for assessing the N transformations in the environment. To fully exploit these tracers, the N and O isotope effects (15ε and 18ε) associated with the respective nitrogen transformation processes must be known. However, the N and O isotope effects of anaerobic ammonium oxidation (anammox), one of the major fixed N sinks and NO3- producers, are not well known. Here, we report the dual N and O isotope effects associated with anammox by three different anammox bacteria including "Ca. Scalindua japonica", a putative marine species, which were measured in continuous enrichment culture experiments. All three anammox species yielded similar N isotope effects of NH4+ oxidation to N2 (15εNH4→N2) ranging from 30.9‰ to 32.7‰ and inverse kinetic isotope effects of NO2- oxidation to NO3- (15εNO2→NO3 = -45.3‰ to -30.1‰). In contrast, 15εNO2→N2 (NO2- reduction to N2) were significantly different among three species, which is probably because individual anammox bacteria species might possess different types of nitrite reductase. We also report the combined O isotope effects for NO2- oxidation (18ENO2→NO3) by anammox bacteria. These obtained dual N and O isotopic effects could provide significant insights into the contribution of anammox bacteria to the fixed N loss and NO2- reoxidation (N recycling) in various natural environments.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Indophenol Blue, Dye content 60 %