Merck
Blockade of nitric oxide synthesis by tyrosine kinase inhibitors in neurones.
Neuropharmacology (1994-11-01)
J Rodriguez, J F Quignard, L Fagni, M Lafon-Cazal, J Bockaert
RESUMEN
In striatal neurones in culture, N-methyl-D-aspartate-(NMDA), kainate-(Kai) and K(+)-dependent cGMP production is entirely mediated via nitric oxide (NO). Low concentrations of lavendustin-A (< or = 0.3 microM), a highly specific tyrosine kinase inhibitor, reduced irreversibly and in a time-dependent manner NMDA-stimulated cGMP production. After a preincubation period of 20 min with lavendustin-A (0.3 microM), the inhibition of NMDA-induced cGMP production was equal to 56 +/- 8% (n = 6). After the same preincubation period, the IC50 of the lavendustin-A blockade was 30 +/- 15 nM. Genistein, another tyrosine kinase inhibitor also inhibited NMDA-dependent cGMP production with high potencies (< or = 3 microM). Whatever the tyrosine kinase inhibitor tested, the basal cGMP production remained unaffected. Kai-, K(+)-, and ionomycin-induced cGMP production was also inhibited by lavendustin-A, and genistein. In contrast, tyrosine kinase inhibitors were unable to block NO donor-induced cGMP production. Using patch clamp experiments, we have also found that lavendustin-A (0.3-1 microM), the most potent tyrosine kinase inhibitor used, (a) did not reduce the NMDA receptor-mediated current, (b) only slighly affected Kai receptor-mediated current (16.4 +/- 3.4% inhibition) and (c) had a marked effect on voltage-sensitive Ca2+ channel- (VSCC) mediated currents (44.4 +/- 4.9% inhibition). A reduction in VSCC activity certainly explains the inhibition of K(+)-, Kai- and possibly part of the NMDA-induced cGMP production.(ABSTRACT TRUNCATED AT 250 WORDS)
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Lavendustin A, A cell-permeable, reversible, and substrate competitive inhibitor of EGF receptor tyrosine kinase and p60c-src with little effect on protein kinase A or protein kinase C.