Merck
Plasmon-induced charge separation at the interface between ITO nanoparticles and TiO2 under near-infrared irradiation.
Physical chemistry chemical physics : PCCP (2019-02-26)
Seung Hyuk Lee, Hiroyasu Nishi, Tetsu Tatsuma
RESUMEN
Plasmon-induced charge separation (PICS) by continuous electron injection from plasmonic compound nanoparticles to a semiconductor was achieved by using solid-state cells based on tin-doped indium oxide (ITO) nanoparticles with a short capping agent and a TiO2 film. The cells extended the PICS range to longer wavelengths and exhibited photoresponses to 1500-2200 nm near-infrared light.