Merck
Linking Herbicide Dissipation to Soil Ecological Risk along Transmission Rights-of-Way in the Yukon Territory, Canada.
Journal of environmental quality (2018-12-05)
Amy Jimmo, Krystal M Isbister, Eric G Lamb, Steven D Siciliano, Katherine J Stewart
RESUMEN
In the Yukon Territory, transmission rights-of-way (ROWs) are managed using brushing and mowing techniques alone. When cut, target species such as Michx. and spp. grow rapidly shortening maintenance cycles. Long-term vegetation control may be improved by integrating herbicide application. However, prior to implementation, the dissipation and toxicity of herbicides in northern latitudes needed to be assessed. The dissipation of Garlon XRT (triclopyr) and Arsenal Powerline (imazapyr) in soils was assessed at five ROW locations representative of the main ecoregion types where ROWs occur within the Yukon Territory. Soils from four sites were collected at 1, 30, and 365 d after treatment to determine persistence of herbicides for each of three application methods (backpack spraying, cut stump, and point injection). Increased sampling intervals were added to better determine the dissipation rate of each herbicide in Yukon Territory soils. Soil dissipation data were linked to a series of standardized toxicity tests, including three soil invertebrates (, , and ). Additionally, the dissipation of both herbicides from the target species L. was assessed at one site. Herbicide residues persisted in soils for longer than 365 d after treatment and longer than 30 d after treatment in . However, concentrations were below the concentration that would affect 25% of the invertebrate species tested. Weight of evidence and toxic exposure ratios were used to characterize the risks associated with herbicide application in northern latitudes and provided both qualitative and quantitative means to communicate the results to the public.