Merck
Severe Hepatotoxicity of Mithramycin Therapy Caused by Altered Expression of Hepatocellular Bile Transporters.
Molecular pharmacology (2019-06-09)
Tristan M Sissung, Phoebe A Huang, Ralph J Hauke, Edel M McCrea, Cody J Peer, Roberto H Barbier, Jonathan D Strope, Ariel M Ley, Mary Zhang, Julie A Hong, David Venzon, Jonathan P Jackson, Kenneth R Brouwer, Patrick Grohar, Jon Glod, Brigitte C Widemann, Theo Heller, David S Schrump, William D Figg
RESUMEN
Mithramycin demonstrates preclinical anticancer activity, but its therapeutic dose is limited by the development of hepatotoxicity that remains poorly characterized. A pharmacogenomics characterization of mithramycin-induced transaminitis revealed that hepatotoxicity is associated with germline variants in genes involved in bile disposition: ABCB4 (multidrug resistance 3) rs2302387 and ABCB11 [bile salt export pump (BSEP)] rs4668115 reduce transporter expression (P < 0.05) and were associated with ≥grade 3 transaminitis developing 24 hours after the third infusion of mithramycin (25 mcg/kg, 6 hours/infusion, every day ×7, every 28 days; P < 0.0040). A similar relationship was observed in a pediatric cohort. We therefore undertook to characterize the mechanism of mithramycin-induced acute transaminitis. As mithramycin affects cellular response to bile acid treatment by altering the expression of multiple bile transporters (e.g., ABCB4, ABCB11, sodium/taurocholate cotransporting polypeptide, organic solute transporter α/β) in several cell lines [Huh7, HepaRG, HepaRG BSEP (-/-)] and primary human hepatocytes, we hypothesized that mithramycin inhibited bile-mediated activation of the farnesoid X receptor (FXR). FXR was downregulated in all hepatocyte cell lines and primary human hepatocytes (P < 0.0001), and mithramycin inhibited chenodeoxycholic acid- and GW4046-induced FXR-galactose-induced gene 4 luciferase reporter activity (P < 0.001). Mithramycin promoted glycochenodeoxycholic acid-induced cytotoxicity in ABCB11 (-/-) cells and increased the overall intracellular concentration of bile acids in primary human hepatocytes grown in sandwich culture (P < 0.01). Mithramycin is a FXR expression and FXR transactivation inhibitor that inhibits bile flow and potentiates bile-induced cellular toxicity, particularly in cells with low ABCB11 function. These results suggest that mithramycin causes hepatotoxicity through derangement of bile acid disposition; results also suggest that pharmacogenomic markers may be useful to identify patients who may tolerate higher mithramycin doses. SIGNIFICANCE STATEMENT: The present study characterizes a novel mechanism of drug-induced hepatotoxicity in which mithramycin not only alters farnesoid X receptor (FXR) and small heterodimer partner gene expression but also inhibits bile acid binding to FXR, resulting in deregulation of cellular bile homeostasis. Two novel single-nucleotide polymorphisms in bile flow transporters are associated with mithramycin-induced liver function test elevations, and the present results are the rationale for a genotype-directed clinical trial using mithramycin in patients with thoracic malignancies.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
ANTI-OST ALPHA(C-TERMINAL) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-SLC51B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution