Merck
  • Prominent role of the Ig-like V domain in trans-interactions of nectins. Nectin3 and nectin 4 bind to the predicted C-C'-C"-D beta-strands of the nectin1 V domain.
Prominent role of the Ig-like V domain in trans-interactions of nectins. Nectin3 and nectin 4 bind to the predicted C-C'-C"-D beta-strands of the nectin1 V domain.
The Journal of biological chemistry (2002-05-16)
Stephanie Fabre, Nicolas Reymond, Francesca Cocchi, Laura Menotti, Patrice Dubreuil, Gabriella Campadelli-Fiume, Marc Lopez
RESUMEN
Nectins form a family of integral molecules that belong to the immunoglobulin superfamily. Their ectodomain is made of three Ig-like domains (V, C, C). This family comprises at least five members, namely nectin1, -2, -3, -4, and poliovirus receptor (PVR), that are involved in different physiological and pathological processes. (i) Nectins are adhesion molecules localized at adherens junctions in epithelial cells. (ii) Some nectins act as poliovirus or alpha-herpesvirus receptors (nectin1). (iii) Nectin1 mutations are involved in orofacial developmental abnormalities in humans. Adhesion properties of nectins are mediated by Ca(2+)-independent homophilic and heterophilic processes through ectodomain trans-interactions. We have described a nectin trans-hetero-interaction network: nectin3 binds to nectin1, nectin2, and PVR; nectin1 also binds to nectin4. In the present study we compared the affinities of the different trans-interactions mediated by nectin1. We found that the K(D) of nectin1/nectin3 and nectin1/nectin4 interactions is 1 and 100 nm, respectively, whereas the K(D) of the nectin1-mediated homophilic interaction is 1 microm. We show that nectin1/nectin3 and nectin1/nectin4 trans-hetero-interactions were mediated through trans V to V domain interactions, whereas C domains contributed to increase the affinity of the interaction. Nectin3 and nectin4 share a common binding region in the nectin1 V domain: (i) nectin3 strongly competed with nectin4 binding, (ii) nectin3 and nectin4 binding to nectin1 was reduced by a number of monoclonal antibodies directed against the nectin1 V domain, and (iii) the glycoprotein D of herpes simplex virus-1 that binds to the V domain of nectin1 reduced nectin3 and nectin4 binding. Finally, using chimeric nectin1/PVR receptors where PVR V domain beta-strands were substituted with the corresponding regions of nectin1, the nectin3 and nectin4 minimal binding region on nectin1 V domain was mapped to the C-C'-C"-D beta-strands.