Merck
Evaluation of dot-blot test for serological diagnosis of bovine brucellosis.
Brazilian journal of microbiology : [publication of the Brazilian Society for Microbiology] (2018-02-18)
Carla Resende Bastos, Luis Antonio Mathias, Márcia Mariza Gomes Jusi, Renata Ferreira Dos Santos, Glaucenyra Cecília Pinheiro da Silva, Marcos Rogério André, Rosangela Zacarias Machado, Karina Paes Bürger
RESUMEN
The objective of this study was to standardize and validate the dot-blot test for the serological diagnosis of bovine brucellosis, compare the results with those found in the 2-mercaptoethanol (2-ME) and complement fixation test (CF), and estimate the relative sensitivity and specificity of the dot-blot compared to these tests. Fifty bovine blood serum samples were used for the test standardization, and 1315 samples were used for evaluation and comparison between the tests; the results were compared using the Kappa indicator. At the end of standardization, it was established as optimal for the antigen obtained from Brucella abortus B19 after passing through a microorganism rupture process, the blood serum samples diluted at 1:100, and the conjugate at 1:30,000. The comparison of the dot-blot results with 2-ME showed Kappa index of 0.9939, sensitivity of 99.48%, and specificity 99.91%, with CF, Kappa index of 0.8226, sensitivity 100% and specificity 95.32%. Using the combination of the test results 2-ME and CF to establish the true condition of the animal, the dot-blot showed relative sensitivity of 100%, and relative specificity of 99.91%. The evaluated test proved to be effective and reliable, besides being easy to handle and interpret the results.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-Bovine IgG (whole molecule)−Alkaline Phosphatase antibody produced in rabbit, affinity isolated antibody