Merck
  • Blockade of the extracellular signal-regulated kinase pathway enhances the therapeutic efficacy of microtubule-destabilizing agents in human tumor xenograft models.
Blockade of the extracellular signal-regulated kinase pathway enhances the therapeutic efficacy of microtubule-destabilizing agents in human tumor xenograft models.
Clinical cancer research : an official journal of the American Association for Cancer Research (2010-02-11)
Kazushi Watanabe, Susumu Tanimura, Aya Uchiyama, Toshiaki Sakamoto, Takumi Kawabata, Kei-ichi Ozaki, Michiaki Kohno
RESUMEN
The extracellular signal-regulated kinase (ERK) pathway is upregulated in human cancers and represents a target for mechanism-based approaches to cancer treatment. However, specific blockade of the ERK pathway alone induces mostly cytostatic rather than proapoptotic effects, resulting in a limited therapeutic efficacy of inhibitors that target the mitogen-activated protein kinase/ERK kinase (MEK). Given the cytoprotective role of the ERK pathway, we examined whether its blockade by the MEK inhibitor PD184352 might enhance the therapeutic efficacy of anticancer drugs in human tumor xenograft models. We recently showed that blockade of the ERK pathway by MEK inhibitors enhances the induction of apoptosis by microtubule-destabilizing agents, including TZT-1027 and vinorelbine, in various tumor cells with aberrant activation of the ERK pathway in vitro. We here examined the therapeutic efficacy of the combination of PD184352 with TZT-1027 or vinorelbine in nude mice harboring HT-29 or HT1080 tumor xenografts, in which the ERK pathway is activated as a result of mutations of BRAF and NRAS, respectively. Coadministration of PD184352 markedly sensitized HT-29 or HT1080 tumor xenografts to TZT-1027-induced or vinorelbine-induced cytotoxicity. Low doses of TZT-1027 or vinorelbine that by themselves showed little or moderate cytotoxicity thus suppressed the growth of HT-29 xenografts almost completely and induced essentially complete regression of HT1080 xenografts when administered with PD184352. The enhanced therapeutic efficacy of the drug combinations was achieved by a relatively transient blockade of the ERK pathway. Administration of both a MEK inhibitor and a microtubule-destabilizing agent represents a promising chemotherapeutic strategy with improved safety for cancer patients.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
PD184352, ≥98% (HPLC)