Authenticated Pancreatic Cancer Cell Lines
In developed countries, pancreatic cancer is one of the leading causes of mortality due to cancer. A 2012 GLOBOCAN estimate attributed 331,000 deaths annually to pancreatic cancer. Subclinical symptoms in the early stages of disease often leads to delayed diagnosis and a poor prognosis, with an estimated 5-year survival rate of less than 5%. If outcomes are not improved, pancreatic cancer is projected to become the second leading cause of cancer-related death in next decade.
Types of pancreatic cancer
Pancreatic cancer may originate from either the exocrine or endocrine compartment of the pancreas. The table below delineates differences between exocrine and endocrine tumors, and will aid in the selection of the appropriate cell line for your research.
Exocrine tumors | Endocrine tumors |
---|---|
|
|
Cell lines: PANC-1, PSN1, AR42J, HaP-T1, HUP-T3, HUP-T4, TGP49, DSL6A/C1, AsPC-1, CFPAC-1 | Cell lines: CRI-G5, CRI-D11, CRI-D2, TGP52, TGP54, TGP55, RIN-M, RIN-5F |
Risk factors
Age is the major risk factor for pancreatic cancer; patients above 50 years of age are at higher risk. Lifestyle factors include smoking, obesity, low physical activity. Diabetes mellitus can be both a risk factor and a consequence of early-stage pancreatic cancer.
Mutations
Approximately 10% of pancreatic cancer patients have a family history of cancer; individuals with mutations in KRAS, TP53, GNAS, SMAD4, CDKN2A, RNF43, ARID1A, and KMT2C may be at higher risk.
Choose cell lines from the table below based on mutation, and click genes to find relevant products (antibodies, shRNA, siRNA, primers, CRISPR plasmids) for your research study.
Mutated gene | Cell line |
---|---|
KRAS | PANC-1, HuP-T3, MIA PaCa-2, AsPC-1, BxPC-3, CFPAC-1, HuP-T4, PSN-1 |
TP53 | PANC-1, HuP-T3, MIA PaCa-2, AsPC-1, BxPC-3, CFPAC-1, HuP-T4, PSN-1 |
GNAS | AsPC-1 |
SMAD4 | AsPC-1 |
CDKN2A | AsPC-1 |
RNF43 | AsPC-1, BxPC-3 |
ARID1A | MIA PaCa-2 |
KMT2C | MIA PaCa-2 |
Small molecules/monoclonal antibodies
Small molecule compounds and antibodies can be used to target cancer cells and block tumor growth and progression. There are small molecule compounds that can target pancreatic cancer based on type of cancer. Drugs used to target pancreatic cancer include:
- Erlotinib
- Capecitabine
- Fluorouracil
- Gemcitabine
- Leucovorin
- Oxaliplatin
- Irinotecan
- Nab-paclitaxel
Applications
Cancer cell lines are indispensable for cancer research, as they provide an accessible, cost-effective model for cellular behavior and response. Based on the characteristics of the cell origins and the experimental need, cell lines may be used in one or more applications. Some examples of application-specific cell line use are included below.
Application | Cell line used |
---|---|
In vitro screening | CFPAC-1, BxPC-3, AsPC-1, PANC-1 and MIA PaCa-2 cell lines were used to screen the anticancer properties of drug conjugates to treat pancreatic cancer1 |
Xenograft models for cancer | PANC-1 cell line was used to generate a xenograft model to evaluate the efficacy of microRNA-delivering nanovectors2 |
Evaluation of novel targets | BxPC-3, MIA PaCa-2 cell line models were used to evaluate mTORC2/RICTOR as a novel target for treatment of human PDAC3 |
Drug response | AsPC-1 pancreatic cancer cell line was employed to investigate the anticancer activity of chemical compounds like manzamine A4 |
Novel therapeutic strategy | Pancreatic cancer cell lines like HuP-T1, HuP-T3 and PANC-1 were used to suggest anti-sense therapy for the K-ras point mutation frequently observed in pancreatic cancers5 |
Toxicity studies | Effects of lipotoxicity studied in insulin-secreting human pancreatic β-cell line, 1.1B46 |
miRNA regulation studies | Role of miR-21 in the pathogenesis of pancreatic ductal adenocarcinoma studied in MIA PaCa-2 cell line7 |
Cell migration studies | Hup-T4 cell lines were used to investigate the role of the ARP2/3 complex in pancreatic cancer cell migration8 |
Network error: Failed to fetch