HPLC Analysis of Peroxidase on TSKgel® G3000SWXL
![HPLC Analysis of Peroxidase on TSKgel® G3000SWXL application for HPLC](/deepweb/assets/sigmaaldrich/product/images/960/414/fe444b23-68a2-4f2e-9422-fd539f394d3e/800/fe444b23-68a2-4f2e-9422-fd539f394d3e.jpg)
Materials
Application
Product No.
Description
Pricing
CONDITIONS
column
TSKgel G3000SWXL, 30 cm × 7.8 mm I.D., 5 μm particles (808541)
mobile phase
0.3M sodium chloride/0.05M phosphate buffer, pH 7.0
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 220 nm
injection
100 μL crude peroxidase preparation from Japanese horseradish (0.15mg)
Description
Legal Information
TSKgel is a registered trademark of Tosoh Corporation