Merck
HPLC Analysis of Nucleosides on Ascentis® Phenyl
HPLC Analysis of Nucleosides on Ascentis® Phenyl application for HPLC
Materials
standard
Product No.
Description
Pricing
Nucleosides Test Mix
in 1% sodium formate (varied conc.), analytical standard
Adenosine
Pharmaceutical Secondary Standard; Certified Reference Material
Disodium Guanylate
Pharmaceutical Secondary Standard; Certified Reference Material
Uridine
Pharmaceutical Secondary Standard; Certified Reference Material
Guanosine
Pharmaceutical Secondary Standard; Certified Reference Material
5′-Adenylic Acid
Pharmaceutical Secondary Standard; Certified Reference Material
Zidovudine Related Compound D
pharmaceutical secondary standard
Cytidine
certified reference material, pharmaceutical secondary standard
CONDITIONS
column
Ascentis Phenyl, 15 cm x 2.1 mm I.D., 5 μm particles (581613-U)
mobile phase
10 mM ammonium formate (pH 3.0 with formic acid)
flow rate
0.2 mL/min
column temp.
35 °C
detector
UV, 270 nm; 750 psi back pressure regulator on outlet of flow cell
injection
1 μL
sample
as indicated, in mobile phase
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany