HPLC Analysis of Aswagandha Extract on Discovery® C18
![HPLC Analysis of Aswagandha Extract on Discovery® C18 application for HPLC](/deepweb/assets/sigmaaldrich/product/images/343/803/ef146935-f357-469a-9477-36fede70783d/800/ef146935-f357-469a-9477-36fede70783d.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Discovery C18, 25 cm x 4.6 mm I.D., 5 μm particles (504971)
mobile phase
[A] 1 mM potassium phosphate, monobasic; [B] acetonitrile
gradient
5 to 45% B in 18 min; 45 to 80% B in 7 min; held at 80% B for 3 min
flow rate
1.5 mL/min
pressure
3191 psi (220 bar)
column temp.
27 °C
detector
UV, 227 nm
injection
20 μL
sample
20 μg/mL in water: acetonitrile (86:14)
Description
Analysis Note
This application demonstrates the suitability of Discovery C18 for the efficient separation of withania extract by the USP monograph.
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany