Merck
HPLC Analysis of 3,5-tBu-MeOBIPHEP Enantiomers on Astec® CHIROBIOTIC® V
HPLC Analysis of 3,5-tBu-MeOBIPHEP Enantiomers on Astec® CHIROBIOTIC® V application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIROBIOTIC® V Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
CONDITIONS
column
CHIROBIOTIC V, 25 cm x 4.6 mm I.D., 5 μm particles (11024AST)
mobile phase
[A] n-hexane; [B] 10% ethanol in n-hexane; (75:25, A:B)
flow rate
1 mL/min
column temp.
18 °C
detector
UV, 230 nm
sample
3,5-tBu-MeOBIPHEP ((6,6′-dimethoxybiphenyl-2,2′-diyl)bis[bis(3,5-di-tert-butylphenyl)phosphine]), 1 mg/mL in ethanol with 5 drops of H2O2 added for oxidation
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC