GC Analysis of Cold Pressed Lemon Essential Oil on SLB®-IL60
Materials
standard
Product No.
Description
Pricing
CONDITIONS
column
SLB-IL60, 30 m x 0.25 mm I.D., 0.25 μm (29505-U)
oven
75 °C (4 min), then 4 °C/min to 200 °C (10 min)
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 25cm/ sec set at 110°C
injection
0.1 μL 100:1 split
liner
4 mm I.D. cup injector
sample
Cold Pressed Lemon Oil, (Sigma), neat
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany