GC Analysis of Volatiles in Water on Equity-1 after SPME using 30 μm PDMS Fiber
Materials
SPME fiber
Product No.
Description
Pricing
SPME fiber assembly Polydimethylsiloxane (PDMS)
df 30 μm(PDMS, needle size 24 ga, for use with autosamplerCONDITIONS
sample/matrix
0.7 mL water spiked with 2-600 μg/liter each analyte, 0.25 g NaCl added
SPME fiber
30 μm PDMS (57309)
extraction
headspace, 12 sec (no stirring), ambient temp.
desorption process
2-3 min, 250 °C (0.75-1.0 mm I.D. inj. port liner)
column
Equity-1, 10 m × 0.20 mm I.D., 1.2 μm (available on request)
oven
70 °C (0.2 min) to 180 °C at 50 °C/min
detector
FID, 250 °C
Description
Legal Information
null