Merck
GC Analysis of Volatiles in Limoncello on SLB®-5ms after SPME using a 100 μm PDMS Fiber
GC Analysis of Volatiles in Limoncello on SLB®-5ms after SPME using a 100 μm PDMS Fiber application for SPME, suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
SPME fiber
Product No.
Description
Pricing
SPME Fiber Assembly
100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, plain red hub
related product
Product No.
Description
Pricing
SPME Fiber Holder
for use with Manual Sampling, pk of 1
SPME Fiber Holder
for use with CTC CombiPAL, Gerstel MPS2 Autosamplers, pk of 1
standard
Product No.
Description
Pricing
Decanal
≥98% (GC), liquid
3,7-Dimethyl-2,6-octadienyl acetate
mixture of isomers, >97.0% (sum of isomers, GC)
(R)-(+)-Limonene
analytical standard
p-Cymene
analytical standard
α-Terpinene
≥95.0% (GC)
γ-Terpinene
analytical standard
Neryl acetate
analytical standard
(+)-α-Pinene
analytical standard
Myrcene
analytical standard
Geraniol
analytical standard
Nonyl acetate
≥97%, FCC
Valencene
natural, ≥65%
Terpinolene
≥95%, FG
Camphene
analytical standard
Nonanal
analytical standard
Linalool
analytical standard
Citral
analytical standard
(−)-α-Bisabolol
analytical standard
Octyl acetate
analytical standard
α-Terpineol
analytical standard
Octanal
analytical standard
α-Phellandrene
natural, ≥85%, FG
Citronellyl acetate
analytical standard
Undecanal
analytical standard
α-Humulene
analytical standard
CONDITIONS
sample/matrix
Limoncello diluted 1:100 in water, then 5 mL of this solution transferred to a 10 mL crimped vial spiked with 100 μL of a 100 ppm solution of nonane (internal standard)
SPME fiber
100 μm polydimethylsiloxane (57300-U)
extraction
headspace, with a pre-equilibration period of 10 min. at 40 °C, and then a fiber exposure of 30 min. at 40 °C, agitated in an alternate clockwise-anticlockwise rotation mode at 500 ppm
desorption process
250 °C for 5 min
column
SLB-5ms, 30 m x 0.25 mm I.D., 0.25 μm (28471-U)
oven
40 °C, 3 °C/min to 250 °C (2 min)
inj. temp.
250 °C
detector
FID, 260 °C (sampling rate 80 ms)
carrier gas
helium, 35.0 cm/sec
injection
splitless
liner
0.75 mm I.D. SPME (2633901)
sample
Limoncello liquor
Description
Analysis Note
In this application, SPME was used for the analysis of volatiles in limoncello using a headspace method. SPME is ideal for the analysis of aroma volatiles in flavored beverages such as liquors. A 100 μm PDMS fiber was inserted into the headspace of the limoncello sample for 30 minutes at 40 °C followed by GC analysis using FID detection. The SLB-5ms GC columns provide consistently low-bleed, inert, efficient, and durable separations.
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany