Merck
GC Analysis of Permanent Gases and C2 Hydrocarbons on a Packed Column (60/80 Carboxen® 1000)
GC Analysis of Permanent Gases and C2 Hydrocarbons on a Packed Column (60/80 Carboxen® 1000) suitable for GC
CONDITIONS
column
15 ft. x 1/8 in. x 2.1 mm I.D., stainless steel
packing
60/80 Carboxen 1000
oven
35 °C (5 min.), 20 °C/min. to 225 °C
detector
TCD
carrier gas
helium, 30 mL/min
injection
0.6 mL, on-column
sample
Scott Gas mix (23437) with added oxygen
Description
Legal Information
Carboxen is a registered trademark of Merck KGaA, Darmstadt, Germany