GC Analysis of Industrial Solvents on Equity-1
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Equity-1, 30 m × 0.32 mm I.D., 1.0 μm (28057-U)
inj. temp.
250 °C
column temp.
35 °C (8 min), 4 °C/min to 130 °C (2 min)
detector
FID, 250 °C
carrier gas
helium, 25 cm/sec @ 35 °C
injection
0.5 μL, 200:1split
liner
split, cup design
sample
59 component neat solvent mixture
Description
Legal Information
null