GC Analysis of Basic Drugs on Equity-5
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Equity-5, 30 m × 0.25 mm I.D., 0.25 μm (28089-U)
oven
45 °C (2 min), 25 °C/min to 110 °C, 15 °C/min to 200 °C, 6 °C/min to 280 °C (3 min)
inj. temp.
250 °C
MSD interface
325 °C
scan range
m/z = 40-450
carrier gas
helium, 40 psi for 0.2 min then 0.7 mL/min constant flow
injection
0.3 μL pulsed splitless @ 50 mL/min (0.5 min)
liner
2 mm I.D., splitless
sample
~15 ng on-column of a 16-component drug standard
Description
Legal Information
null