Merck
  • Characteristics of speech production in patients with T1 glottic cancer who underwent laser cordectomy or radiotherapy.
Characteristics of speech production in patients with T1 glottic cancer who underwent laser cordectomy or radiotherapy.
Logopedics, phoniatrics, vocology (2017-10-05)
Yong Tae Hong, Min Ju Park, Ki Hwan Hong
ABSTRACT
Laser cordectomy (LC) or radiotherapy (RT) is often recommended in the early stage of laryngeal cancer. We conducted perceptual and acoustic analysis to compare sustained vowel and stop consonants since there is no article evaluating both the sustained vowel and stop consonants. Eventually, we might determine which management is superior in terms of speech production. A total of 28 patients who underwent LC and RT for early T1 glottic cancer were selected. The sustained vowel /a/ and bilabial stop consonants were used to assess the perceptual scores. The fundamental frequency (F The GRBAS and discrimination scores were not significantly different between two groups. F The differences may have been due to muscular fibrosis after RT. Movements of vocal cords with fibrosis were sluggish, when impulsion developed to pronounce the initial /p/ sound, so the VOT was shortened and the VD was longer after RT.
Product Description
Supelco
VOCOL® Capillary GC Column, L × I.D. 60 m × 0.53 mm, df 3.00 μm
Supelco
VOCOL® Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 3.00 μm
Supelco
VOCOL® Capillary GC Column, L × I.D. 60 m × 0.25 mm, df 1.50 μm
Supelco
VOCOL® Capillary GC Column, L × I.D. 105 m × 0.53 mm, df 3.00 μm
Supelco
VOCOL® Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 1.50 μm
Supelco
VOCOL® Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 1.80 μm
Supelco
VOCOL® Capillary GC Column, L × I.D. 30 m × 0.53 mm, df 3.00 μm
Supelco
VOCOL® Capillary GC Column, L × I.D. 20 m × 0.18 mm, df 1.00 μm