Merck
  • Process sampling module coupled with purge and trap-GC-FID for in situ auto-monitoring of volatile organic compounds in wastewater.
Process sampling module coupled with purge and trap-GC-FID for in situ auto-monitoring of volatile organic compounds in wastewater.
Talanta (2009-10-20)
Hsin-Wang Liu, Yung-Tsun Liu, Bei-Zen Wu, Hung-Chi Nian, Hsing-Jung Chen, Kong-Hwa Chiu, Jiunn-Guang Lo
ABSTRACT
An automatic sampling device, i.e., process sampling module (PSM), connected with a purge and trap-GC-FID system has been developed for real-time monitoring of VOCs in wastewater. The system was designed to simultaneously monitor 17 compounds, including one polar compound, i.e., acetone, and 16 non-polar compounds. The trapping tube is packed with two adsorbents, Carbopack B and Carbosieve III, to trap target compounds. For the purpose of in situ monitoring, the flush valve of the sampling tube is composed of two two-way valves and a time controller to prevent absorption interference of the residue. The optimal conditions for the analytical system include a 12 min purge time at a temperature of 60 degrees C, and 4 min of desorption time with a desorption temperature of 260 degrees C. Good chromatograms have been obtained with the analytical system even if a cryogenic device and de-misting were not used. The relative standards deviation (RSD) of the system is between 2% and 13.4%, and accuracies between 0.3 and 23.5% have been achieved. The detection limits of the method range from 0.32 to 2.39 ppb. In this system, the four parts, i.e., PSM, P&T, GC, and FID, were simple, reliable and rugged. Also, the interface of these four parts was simple and dependable.
Product Description
Supelco
Carbosieve Adsorbent, matrix Carbosieve S-III, 60-80 mesh, bottle of 10 g
Supelco
Carbosieve Adsorbent, matrix Carbosieve S-II, 60-80 mesh, bottle of 10 g
Supelco
Carbosieve Adsorbent, matrix Carbosieve G, 60-80 mesh, bottle of 5 g
Supelco
Carbosieve Adsorbent, matrix Carbosieve S-II, 80-100 mesh, bottle of 10 g
Supelco
Carbosieve Adsorbent, matrix Carbosieve G, 80-100 mesh, bottle of 5 g
Supelco
Carbopack Adsorbent, matrix Carbopack B, 60-80 mesh, bottle of 10 g
Supelco
Carbosieve SIII, stainless steel TD tube, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea
Supelco
Carbopack X, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
Supelco
Carbopack B, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., unconditioned, pkg of 10 ea, matrix Carbopack
Supelco
Carbopack Adsorbent, matrix Carbopack X, 60-80 mesh, bottle of 10 g
Supelco
Carbopack Adsorbent, matrix Carbopack C, 80-100 mesh, bottle of 10 g
Supelco
Carbopack Adsorbent, matrix Carbopack X, 120-400 mesh, bottle of 50 g
Supelco
Carbopack Adsorbent, matrix Carbopack X, 40-60 mesh, bottle of 10 g
Supelco
Carbopack Adsorbent, matrix Carbopack C, 60-80 mesh, bottle of 10 g
Supelco
Carbopack B, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
Supelco
Carbopack B, stainless steel TD tube, O.D. × L 1/4 × 3 1/2, unconditioned, pkg of 10 ea
Supelco
Carbopack B, O.D. × L 1/4 in. × 3 1/2 in., glass TD tube, fritted, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
Supelco
FLM Carbopack X Deactivated Stainless Steel TD Tube, preconditioned, pkg of 10 tubes, O.D. × L 1/4 in. (6.35 mm) × 3 1/2 in. (89 mm)