Merck
Influenza A virus replication induces cell cycle arrest in G0/G1 phase.
Journal of virology (2010-09-24)
Yuan He, Ke Xu, Bjoern Keiner, Jianfang Zhou, Volker Czudai, Tianxian Li, Ze Chen, Jinhua Liu, Hans-Dieter Klenk, Yue Long Shu, Bing Sun
ABSTRACT
Many viruses interact with the host cell division cycle to favor their own growth. In this study, we examined the ability of influenza A virus to manipulate cell cycle progression. Our results show that influenza A virus A/WSN/33 (H1N1) replication results in G(0)/G(1)-phase accumulation of infected cells and that this accumulation is caused by the prevention of cell cycle entry from G(0)/G(1) phase into S phase. Consistent with the G(0)/G(1)-phase accumulation, the amount of hyperphosphorylated retinoblastoma protein, a necessary active form for cell cycle progression through late G(1) into S phase, decreased after infection with A/WSN/33 (H1N1) virus. In addition, other key molecules in the regulation of the cell cycle, such as p21, cyclin E, and cyclin D1, were also changed and showed a pattern of G(0)/G(1)-phase cell cycle arrest. It is interesting that increased viral protein expression and progeny virus production in cells synchronized in the G(0)/G(1) phase were observed compared to those in either unsynchronized cells or cells synchronized in the G(2)/M phase. G(0)/G(1)-phase cell cycle arrest is likely a common strategy, since the effect was also observed in other strains, such as H3N2, H9N2, PR8 H1N1, and pandemic swine H1N1 viruses. These findings, in all, suggest that influenza A virus may provide favorable conditions for viral protein accumulation and virus production by inducing a G(0)/G(1)-phase cell cycle arrest in infected cells.
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder