Merck
  • Characterization of epithelial-mesenchymal transition intermediate/hybrid phenotypes associated to resistance to EGFR inhibitors in non-small cell lung cancer cell lines.
Characterization of epithelial-mesenchymal transition intermediate/hybrid phenotypes associated to resistance to EGFR inhibitors in non-small cell lung cancer cell lines.
Oncotarget (2017-12-22)
Valentina Fustaino, Dario Presutti, Teresa Colombo, Beatrice Cardinali, Giuliana Papoff, Rossella Brandi, Paola Bertolazzi, Giovanni Felici, Giovina Ruberti
ABSTRACT
Increasing evidence points to a key role played by epithelial-mesenchymal transition (EMT) in cancer progression and drug resistance. In this study, we used
Sigma-Aldrich
Erlotinib hydrochloride
Sigma-Aldrich
LY-364947, ≥98% (HPLC)