Merck
  • Snail Is a Direct Target of Hypoxia-inducible Factor 1α (HIF1α) in Hypoxia-induced Endothelial to Mesenchymal Transition of Human Coronary Endothelial Cells.
Snail Is a Direct Target of Hypoxia-inducible Factor 1α (HIF1α) in Hypoxia-induced Endothelial to Mesenchymal Transition of Human Coronary Endothelial Cells.
The Journal of biological chemistry (2015-05-15)
Xingbo Xu, Xiaoying Tan, Björn Tampe, Elisa Sanchez, Michael Zeisberg, Elisabeth M Zeisberg
ABSTRACT
Endothelial to mesenchymal transition (EndMT) was originally described in heart development where the endocardial endothelial cells that line the atrioventricular canal undergo an EndMT to form the endocardial mesenchymal cushion that later gives rise to the septum and mitral and tricuspid valves. In the postnatal heart specifically, endothelial cells that originate from the endocardium maintain increased susceptibility to undergo EndMT as remnants from their embryonic origin. Such EndMT involving adult coronary endothelial cells contributes to microvascular rarefaction and subsequent chronification of hypoxia in the injured heart, ultimately leading to cardiac fibrosis. Although in most endothelial beds hypoxia induces tip cell formation and sprouting angiogenesis, here we demonstrate that hypoxia is a stimulus for human coronary endothelial cells to undergo phenotypic changes reminiscent of EndMT via a mechanism involving hypoxia-inducible factor 1α-induced activation of the EndMT master regulatory transcription factor SNAIL. Our study adds further evidence for the unique susceptibility of endocardium-derived endothelial cells to undergo EndMT and provides novel insights into how hypoxia contributes to progression of cardiac fibrosis. Additional studies may be required to discriminate between distinct sprouting angiogenesis and EndMT responses of different endothelial cells populations.
Sigma-Aldrich
Anti-S100A4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone B-5-1-2, ascites fluid