Merck
  • Lunatic fringe-mediated Notch signaling regulates adult hippocampal neural stem cell maintenance.
Lunatic fringe-mediated Notch signaling regulates adult hippocampal neural stem cell maintenance.
eLife (2017-07-13)
Fatih Semerci, William Tin-Shing Choi, Aleksandar Bajic, Aarohi Thakkar, Juan Manuel Encinas, Frederic Depreux, Neil Segil, Andrew K Groves, Mirjana Maletic-Savatic
ABSTRACT
Hippocampal neural stem cells (NSCs) integrate inputs from multiple sources to balance quiescence and activation. Notch signaling plays a key role during this process. Here, we report that Lunatic fringe (Lfng), a key modifier of the Notch receptor, is selectively expressed in NSCs. Further, Lfng in NSCs and Notch ligands Delta1 and Jagged1, expressed by their progeny, together influence NSC recruitment, cell cycle duration, and terminal fate. We propose a new model in which Lfng-mediated Notch signaling enables direct communication between a NSC and its descendants, so that progeny can send feedback signals to the 'mother' cell to modify its cell cycle status. Lfng-mediated Notch signaling appears to be a key factor governing NSC quiescence, division, and fate.
Sigma-Aldrich
Anti-Polysialic Acid-NCAM Antibody, clone 2-2B, ascites fluid, clone 2-2B, Chemicon®
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP) antibody produced in mouse, clone G-A-5, ascites fluid
Sigma-Aldrich
Anti-HES-5 Antibody, Chemicon®, from rabbit