Merck
  • Halogenated fatty amides - A brand new class of disinfection by-products.
Halogenated fatty amides - A brand new class of disinfection by-products.
Water research (2017-10-20)
Dmitry S Kosyakov, Nikolay V Ul'yanovskii, Mark S Popov, Tomas B Latkin, Albert T Lebedev
ABSTRACT
An array of similar halogenated nitrogen-containing compounds with elemental composition CnH2nNO2X, CnH2n-2NO2X and CnH2n-1NOX2 (X = Cl, Br; n = 16, 18, 22) was detected in drinking water with high performance liquid chromatography - high resolution mass spectrometry (HPLC-HRMS) method. Compounds of this type were never mentioned among disinfection by-products. Tandem mass spectrometry allowed referring them to halohydrines or dihalogenated fatty amides, the products of conjugated electrophilic addition of halogens to the double bonds of unsaturated fatty amides. The proposed structures were confirmed by conducting aqueous chlorination with standard solution of oleamide. These compounds may be considered as a brand new class of disinfection by products, while their toxicities require special study.
Sigma-Aldrich
Oleamide, ≥99%
Milli-Q
LC-Pak® Polisher, Placed at the point of dispense of Milli-Q® IQ/EQ 7 series water systems. Suitable for HPLC, UHPLC, LC-MS and LC-MS/MS analyses.