Merck
How ingredients influence furan and aroma generation in sponge cake.
Food chemistry (2017-12-31)
Mayela Cepeda-Vázquez, Barbara Rega, Nicolas Descharles, Valérie Camel
ABSTRACT
A wide range of compounds can be formed during thermal processing of food, some of which are relevant for aroma (e.g., furfural), while others are of great health concern (e.g., furan). This paper presents the study of formulation as affecting the simultaneous generation of furan and furfural, along with other aroma quality markers, in sponge cake by means of headspace trap/GC-MS. Ingredients were screened according to their category (fat, salt, sugar, egg-based). Glucose-containing formulation resulted in the highest content of furan and furfural (12.5 ± 0.5 ng g
Sigma-Aldrich
Hydroxyacetone, contains ≤500 ppm sodium carbonate as stabilizer, technical grade, 90%
Sigma-Aldrich
Furan-d4, ≥98 atom % D, ≥99% (CP), contains 0.025 wt. % BHT as stabilizer