Merck
  • Evidence of hydrocarbon nanodrops in highly ordered stratum corneum model membranes.
Evidence of hydrocarbon nanodrops in highly ordered stratum corneum model membranes.
Journal of lipid research (2017-11-03)
Adrian Paz Ramos, Gert Gooris, Joke Bouwstra, Michel Lafleur
ABSTRACT
The stratum corneum (SC), the top layer of skin, dictates the rate of both water loss through the skin and absorption of exogenous molecules into the body. The crystalline organization of the lipids in the SC is believed to be a key feature associated with the very limited permeability of the skin. In this work, we characterized the organization of SC lipid models that include, as in native SC, cholesterol, a series of FFAs (saturated with C16-C24 chains), as well as a ceramide bearing an oleate chain-linked to a very long saturated acyl chain [
Sigma-Aldrich
Lignoceric acid, ≥99% (GC)