Merck
  • Inhibition of EZH2 triggers the tumor suppressive miR-29b network in multiple myeloma.
Inhibition of EZH2 triggers the tumor suppressive miR-29b network in multiple myeloma.
Oncotarget (2018-01-02)
Maria Angelica Stamato, Giada Juli, Enrica Romeo, Domenica Ronchetti, Mariamena Arbitrio, Daniele Caracciolo, Antonino Neri, Pierosandro Tagliaferri, Pierfrancesco Tassone, Nicola Amodio
ABSTRACT
Downregulation of tumor suppressor (TS) microRNAs (miRNAs) commonly occurs in human cancer, including multiple myeloma (MM). We previously demonstrated that miR-29b is a relevant TS miRNA, whose expression in MM cells is inhibited by HDAC4-dependent deacetylation. Here, we provide novel insights into epigenetic mechanisms suppressing miR-29b in MM. In MM patient-derived plasma cells, we found inverse correlation between miR-29b and EZH2 mRNA expression. Both siRNAs and pharmacologic inhibitors of EZH2 led to miR-29b upregulation, and this effect was ascribed to reduced H3K27-trimethylation (H3K27me3) of miR-29a/b-1 promoter regions. Induction of miR-29b upon EZH2 inhibition occurred together with downregulation of major miR-29b pro-survival targets, such as SP1, MCL-1 and CDK6. Knock-down of the EZH2-interacting long non-coding RNA MALAT1 also reduced H3K27me3 of miR-29a/b-1 promoter, along with induction of miR-29b and downregulation of miR-29b targets. Importantly, inhibition of miR-29b by antagomiRs dramatically reduced
Sigma-Aldrich
MISSION® esiRNA, targeting human EZH2
Sigma-Aldrich
GSK343, ≥98% (HPLC)
Sigma-Aldrich
3-Deazaneplanocin A hydrochloride, ≥97% (HPLC)