Merck
  • Photochemical delivery of bleomycin induces T-cell activation of importance for curative effect and systemic anti-tumor immunity.
Photochemical delivery of bleomycin induces T-cell activation of importance for curative effect and systemic anti-tumor immunity.
Journal of controlled release : official journal of the Controlled Release Society (2017-10-19)
Ole-Jacob Norum, Ane Sofie Viset Fremstedal, Anette Weyergang, Jakub Golab, Kristian Berg
ABSTRACT
Photochemical internalization (PCI) is a technology to enhance intracellular drug delivery by light-induced translocation of endocytosed therapeutics into the cytosol. The aim of this study was to explore the efficacy of PCI-based delivery of bleomycin and the impact on systemic anti-tumor immunity. Mouse colon carcinoma cells (CT26.CL25), stably expressing the bacterial β-galactosidase, were inoculated into the legs of athymic or immuno-competent BALB/c mice strains. The mice were injected with the photosensitizer AlPcS
Sigma-Aldrich
29H,31H-Phthalocyanine, β-form, 98%