Merck
  • Zinc strengthens the jejunal barrier by reversibly tightening the paracellular route.
Zinc strengthens the jejunal barrier by reversibly tightening the paracellular route.
American journal of physiology. Gastrointestinal and liver physiology (2017-09-03)
Silke S Zakrzewski, Michael Fromm, Jörg D Schulzke, Dorothee Günzel
ABSTRACT
During the postweaning period, piglets are prone to gastrointestinal infections. The resulting impairment of intestinal barrier function may cause diarrhea associated with growth retardation or even death of piglets. Orally applied Zn is commonly used to prevent and treat diarrhea, but its mode of action still needs to be elucidated. To analyze the molecular mechanism whereby Zn acts on porcine intestinal barrier function, ex vivo studies on piglet jejunum and accompanying in vitro studies on a porcine jejunal epithelial cell line, IPEC-J2/PS, were performed with electrophysiological tools. Feeding pharmacological Zn doses exerted no significant electrophysiologically ascertainable short- and long-term effects on jejunal barrier function ex vivo. However, in IPEC-J2/PS, basolateral Zn was cytotoxic since its application caused a release of lactate dehydrogenase and an irreversible breakdown of the epithelial barrier. In contrast, apical Zn application caused an immediate increase in paracellular resistance and a decrease in permeability to the paracellular marker fluorescein, reflecting overall barrier strengthening in vitro. Apical effects were fully reversible upon washout. This indicates that Zn supplemented to feed was completely washed out during ex vivo jejunum preparation. We conclude that there is no evidence for long-term barrier effects through prophylactic Zn supplementation and that extracellular Zn acts acutely and reversibly from the apical side via tightening the paracellular route, thus counteracting leak-flux diarrhea.
Product Description
Selenium, powder, 250 max. part. size (micron), purity 99.95%, weight 100 g
Sigma-Aldrich
AIDA, solid
Sigma-Aldrich
MEM Non-essential Amino Acid Solution (100×), without L-glutamine, liquid, sterile-filtered, BioReagent, suitable for cell culture