Merck
  • Human Semen Quality, Sperm DNA Damage, and the Level of Reproductive Hormones in Relation to Urinary Concentrations of Parabens.
Human Semen Quality, Sperm DNA Damage, and the Level of Reproductive Hormones in Relation to Urinary Concentrations of Parabens.
Journal of occupational and environmental medicine (2017-07-12)
Joanna Jurewicz, Michał Radwan, Bartosz Wielgomas, Emila Dziewirska, Anetta Karwacka, Anna Klimowska, Paweł Kałużny, Paweł Radwan, Michał Bochenek, Wojciech Hanke
ABSTRACT
The aim of this study was to evaluate the association between environmental exposure to parabens and semen quality parameters [main semen parameters, computer-aided semen analysis (CASA parameters], sperm chromatin structure, and the level of reproductive hormones in men [follicle-stimulating hormone (FSH), testosterone, estradiol]. Urine samples collected from 315 men who attended the infertility clinic for diagnostic purposes with normal semen concentration of 15 to 300 mln/mL were analyzed for five parabens concentrations using a validated gas chromatography ion-tap mass spectrometry method. Participants were interviewed and also provided a semen, saliva, and blood samples. Urinary parabens concentrations were significantly associated with an increase in the percentage of sperm with abnormal morphology, in sperm with high DNA stainability and a decrease in the percentage of motility and testosterone level. This is one of the first study on this topic, so the observation of the relationship between parabens and semen quality warrants further investigation.
Sigma-Aldrich
Isobutyl 4-hydroxybenzoate, 97%