MilliporeSigma
  • Engaging the Terminal: Promoting Halogen Bonding Interactions with Uranyl Oxo Atoms.
Engaging the Terminal: Promoting Halogen Bonding Interactions with Uranyl Oxo Atoms.
Chemistry (Weinheim an der Bergstrasse, Germany) (2017-07-15)
Korey P Carter, Mark Kalaj, Robert G Surbella, Lucas C Ducati, Jochen Autschbach, Christopher L Cahill
ABSTRACT
Engaging the nominally terminal oxo atoms of the linear uranyl (UO
Sigma-Aldrich
4′-Chloro-2,2′:6′,2′′-terpyridine, 99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
1,10-Phenanthroline, ≥99%