Merck
A new role for human dyskerin in vesicular trafficking.
FEBS open bio (2017-10-06)
Nunzia Di Maio, Rosario Vicidomini, Alberto Angrisani, Valentina Belli, Maria Furia, Mimmo Turano
ABSTRACT
Dyskerin is an essential, conserved, multifunctional protein found in the nucleolus, whose loss of function causes the rare genetic diseases X-linked dyskeratosis congenita and Hoyeraal-Hreidarsson syndrome. To further investigate the wide range of dyskerin's biological roles, we set up stable cell lines able to trigger inducible protein knockdown and allow a detailed analysis of the cascade of events occurring within a short time frame. We report that dyskerin depletion quickly induces cytoskeleton remodeling and significant alterations in endocytic Ras-related protein Rab-5A/Rab11 trafficking. These effects arise in different cell lines well before the onset of telomere shortening, which is widely considered the main cause of dyskerin-related diseases. Given that vesicular trafficking affects many homeostatic and differentiative processes, these findings add novel insights into the molecular mechanisms underlining the pleiotropic manifestation of the dyskerin loss-of-function phenotype.
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting human DKC1
Pricing and availability is not currently available.
Sigma-Aldrich
Latrunculin A, from sea sponge, ≥85% (HPLC), waxy solid
Pricing and availability is not currently available.