Merck
  • Characterization and epitope mapping of Dengue virus type 1 specific monoclonal antibodies.
Characterization and epitope mapping of Dengue virus type 1 specific monoclonal antibodies.
Virology journal (2017-10-04)
Wen-Hung Chen, Feng-Pai Chou, Yu-Kuo Wang, Sheng-Cih Huang, Chuan-Hung Cheng, Tung-Kung Wu
ABSTRACT
Dengue virus (DV) infection causes a spectrum of clinical diseases ranging from dengue fever to a life-threatening dengue hemorrhagic fever. Four distinct serotypes (DV1-4), which have similar genome sequences and envelope protein (E protein) antigenic properties, were divided. Among these 4 serotypes, DV1 usually causes predominant infections and fast diagnosis and effective treatments are urgently required to prevent further hospitalization and casualties. To develop antibodies specifically targeting and neutralizing DV1, we immunized mice with UV-inactivated DV1 viral particles and recombinant DV1 E protein from residue 1 to 395 (E395), and then generated 12 anti-E monoclonal antibodies (mAbs) as the candidates for a series of characterized assays such as ELISA, dot blot, immunofluorescence assay, western blot, and foci forming analyses. Among the mAbs, 10 out of 12 showed cross-reactivity to four DV serotypes as well as Japanese encephalitis virus (JEV) in different cross-reactivity patterns. Two particular mAbs, DV1-E1 and DV1-E2, exhibited strong binding specificity and neutralizing activity against DV1 and showed no cross-reactivity to DV2, DV3, DV4 or JEV-infected cells as characterized by ELISA, dot blot, immunofluorescence assay, western blot, and foci forming analyses. Using peptide coated indirect ELISA, we localized the neutralizing determinants of the strongly inhibitory mAbs to a sequence-unique epitope on the later-ridge of domain III of the DV1 E protein, centered near residues T346 and D360 ( Our results showed two selected mAbs DV1-E1 and DV1-E2 can specifically target and significantly neutralize DV1. With further research these two mAbs might be applied in the development of DV1 specific serologic diagnosis and used as a feasible treatment option for DV1 infection. The identification of DV1 mAbs epitope with key residues can also provide vital information for vaccine design.
Sigma-Aldrich
Polyethylene glycol/ dimethyl sulfoxide solution, Hybri-Max, average mol wt 1,450, 50 % (w/v), 0.2 μm filtered, BioReagent, suitable for hybridoma