Merck
  • Structure-activity relationship study of ProxyPhos chemosensors for the detection of proximal phosphorylation and other phosphate species.
Structure-activity relationship study of ProxyPhos chemosensors for the detection of proximal phosphorylation and other phosphate species.
The Analyst (2017-09-21)
A D Cabral, B I Murcar-Evans, K Toutah, M Bancerz, D Rosa, K Yuen, T B Radu, M Ali, A Penkul, D Kraskouskaya, P T Gunning
ABSTRACT
Chemosensors for the detection of phosphate-containing biological species are in high need. Detection of proximally phosphorylated sites of PP
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, 10×, Modified, without calcium chloride and magnesium chloride, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)