Merck
  • Transformation of iodide and formation of iodinated by-products in heat activated persulfate oxidation process.
Transformation of iodide and formation of iodinated by-products in heat activated persulfate oxidation process.
Chemosphere (2017-05-02)
Lu Wang, Deyang Kong, Yuefei Ji, Junhe Lu, Xiaoming Yin, Quansuo Zhou
ABSTRACT
Formation of halogenated disinfection by-products (DBPs) in sulfate radical-based advanced oxidation processes (SR-AOPs) have attracted considerable concerns recently. Previous studies have focused on the formation of chlorinated and brominated DBPs. This research examined the transformation of I
Sigma-Aldrich
4-Iodophenol, 99%
Sigma-Aldrich
2-Iodophenol, 98%