Merck
  • Discovery and characterization of a mammalian amyloid disaggregation activity.
Discovery and characterization of a mammalian amyloid disaggregation activity.
Protein science : a publication of the Protein Society (2010-02-18)
Amber N Murray, James P Solomon, Ya-Juan Wang, William E Balch, Jeffery W Kelly
ABSTRACT
The formation of amyloid, a cross-beta-sheet fibrillar aggregate, is associated with a variety of aging-associated degenerative diseases. Herein, we report the existence of a mammalian amyloid disaggregase activity that is present in all tissues and cell types tested. Homogenates from mammalian tissues and cell lines are able to disaggregate amyloid fibrils composed of amyloid beta (A beta)(1-40) or the 8 kDa plasma gelsolin fragment. The mammalian disaggregase activity is sensitive to proteinase K digestion and can be uncoupled from proteolysis activity using a protease inhibitor cocktail. Amyloid disaggregation and proteolysis activities are remarkably resistant to changes in temperature and pH. Identification and manipulation of the proteins responsible for the amyloid disaggregation/degradation activities offers the possibility of ameliorating aggregation-associated diseases.
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥200 units/mg protein, lyophilized powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Actin Antibody, clone C4, clone C4, Chemicon®, from mouse