Merck
Identification of S-Nitrosylated (SNO) Proteins in
Frontiers in cellular and infection microbiology (2017-06-08)
Meirav Trebicz-Geffen, Preeti Shahi, Shruti Nagaraja, Shai Vanunu, Shiran Manor, Amit Avrahami, Serge Ankri
ABSTRACT
We have recently reported that
Sigma-Aldrich
N-[Tris(hydroxymethyl)methyl]acrylamide, contains ≤7% KCl, 93%