Merck
  • A liquid chromatography-tandem mass spectrometry method to measure fatty acids in biological samples.
A liquid chromatography-tandem mass spectrometry method to measure fatty acids in biological samples.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2017-05-04)
Milene Volpato, Jade A Spencer, Amanda D Race, Alessandra Munarini, Andrea Belluzzi, Andrew J Cockbain, Mark A Hull, Paul M Loadman
ABSTRACT
As pre-clinical and clinical research interest in ω-3 polyunsaturated fatty acids (PUFA) increases, so does the need for a fast, accurate and reproducible analytical method to measure fatty acids (FA) in biological samples in order to validate potential prognostic and predictive biomarkers, as well as establishing compliance in ω-3 PUFA intervention trials. We developed a LC-ESI-MS/MS method suitable for high throughput development to measure FAs and validated it in the context of treatment with the ω-3 PUFA eicosapentaenoic acid (EPA). Uniquely we directly compared the LC-ESI-MS/MS method to a GC-MS protocol. We demonstrated the LC-ESI-MS/MS method is accurate and reproducible, with coefficients of variation consistently below 15% for each PUFA analysed. The relative FA content values correlated well with those obtained by GC-MS (r
Product Description
Supelco
4-[2-(N,N-Dimethylamino)ethylaminosulfonyl]-7-(2-aminoethylamino)-2,1,3-benzoxadiazole, derivatization grade (HPLC), LiChropur, ≥95.0% (HPLC)
Sigma-Aldrich
Benzofurazan, 97%
Sigma-Aldrich
2-(Diisopropylamino)ethyl methacrylate, 97%, contains ~100 ppm monomethyl ether hydroquinone as inhibitor