Merck
  • Genetic, structural, and chemical insights into the dual function of GRASP55 in germ cell Golgi remodeling and JAM-C polarized localization during spermatogenesis.
Genetic, structural, and chemical insights into the dual function of GRASP55 in germ cell Golgi remodeling and JAM-C polarized localization during spermatogenesis.
PLoS genetics (2017-06-16)
Amandine Cartier-Michaud, Anne-Laure Bailly, Stéphane Betzi, Xiaoli Shi, Jean-Claude Lissitzky, Ana Zarubica, Arnauld Sergé, Philippe Roche, Adrien Lugari, Véronique Hamon, Florence Bardin, Carine Derviaux, Frédérique Lembo, Stéphane Audebert, Sylvie Marchetto, Bénédicte Durand, Jean-Paul Borg, Ning Shi, Xavier Morelli, Michel Aurrand-Lions
ABSTRACT
Spermatogenesis is a dynamic process that is regulated by adhesive interactions between germ and Sertoli cells. Germ cells express the Junctional Adhesion Molecule-C (JAM-C, encoded by Jam3), which localizes to germ/Sertoli cell contacts. JAM-C is involved in germ cell polarity and acrosome formation. Using a proteomic approach, we demonstrated that JAM-C interacted with the Golgi reassembly stacking protein of 55 kDa (GRASP55, encoded by Gorasp2) in developing germ cells. Generation and study of Gorasp2-/- mice revealed that knock-out mice suffered from spermatogenesis defects. Acrosome formation and polarized localization of JAM-C in spermatids were altered in Gorasp2-/- mice. In addition, Golgi morphology of spermatocytes was disturbed in Gorasp2-/- mice. Crystal structures of GRASP55 in complex with JAM-C or JAM-B revealed that GRASP55 interacted via PDZ-mediated interactions with JAMs and induced a conformational change in GRASP55 with respect of its free conformation. An in silico pharmacophore approach identified a chemical compound called Graspin that inhibited PDZ-mediated interactions of GRASP55 with JAMs. Treatment of mice with Graspin hampered the polarized localization of JAM-C in spermatids, induced the premature release of spermatids and affected the Golgi morphology of meiotic spermatocytes.
Product Description
Glutathione Sepharose 4B, Cytiva 17-0756-01, pack of 10 mL
Sigma-Aldrich
Anti-Actin antibody, Mouse monoclonal, clone AC-40, purified from hybridoma cell culture
Sigma-Aldrich
Lectin from Arachis hypogaea (peanut), biotin conjugate, lyophilized powder