Merck
  • Upregulation of miR-195 accelerates oxidative stress-induced retinal endothelial cell injury by targeting mitofusin 2 in diabetic rats.
Upregulation of miR-195 accelerates oxidative stress-induced retinal endothelial cell injury by targeting mitofusin 2 in diabetic rats.
Molecular and cellular endocrinology (2017-05-11)
Rui Zhang, Qian Garrett, Huimin Zhou, Xiaoxi Wu, Yueran Mao, Ximing Cui, Bing Xie, Zanchao Liu, Dongsheng Cui, Lei Jiang, Qingfu Zhang, Shunjiang Xu
ABSTRACT
This study was performed to investigate the oxidative stress-induced miRNA changes in relation to pathogenesis of diabetic retinopathy (DR) and to establish a functional link between miRNAs and oxidative stress-induced retinal endothelial cell injury. Our results demonstrated that oxidative stress could induce alterations of miRNA expression profile, including up-regulation of miR-195 in the diabetic retina or cultured HMRECs after exposed to H
Sigma-Aldrich
MISSION® esiRNA, targeting human MFN2
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder