Merck
  • Hepatic Uptake Mechanism of Ophiopogonin D Mediated by Organic Anion Transporting Polypeptides.
Hepatic Uptake Mechanism of Ophiopogonin D Mediated by Organic Anion Transporting Polypeptides.
European journal of drug metabolism and pharmacokinetics (2016-11-07)
Wen Zhang, Xiaomin Xiong, Lin Chen, Mingyi Liu, Yuqing Xiong, Hong Zhang, Shibo Huang, Chunhua Xia
ABSTRACT
Ophiopogonin D (OPD) is one of the main active ingredients of SMI (Shenmai injection) which is widely used in clinical practice in China. Our previous study indicated  that OPD might be transported from blood into liver mediated by organic anion transporting polypeptides (OATPs/oatps). This study aims to explore the hepatic uptake mechanism of OPD in rat and human. Rosuvastatin (a competitive inhibitor of oatp1b2, oatp1a1, and oatp1a4), glycyrrhizic acid (a specific inhibitor of oatp1b2), digoxin (a specific inhibitor of oatp1a4), bromosulfophthalein (BSP), and ibuprofen (a specific inhibitor of oatp1a1) were used to study the uptake of OPD in rat hepatocytes. Furthermore, the uptake of OPD in human OATP1B1*1a-HEK293T cells was also investigated, and rosuvastatin, BSP, rifampin, and glycyrrhizic acid were all used as the competitive inhibitor of OATP1B1. OPD can be taken in rat primary hepatocytes with K Overall, this study indicates OATP1B1 in human and oatp1b2 in rats might participate in the hepatic uptake of OPD.