Merck
  • Diverse roles of guanine nucleotide exchange factors in regulating collective cell migration.
Diverse roles of guanine nucleotide exchange factors in regulating collective cell migration.
The Journal of cell biology (2017-05-18)
Assaf Zaritsky, Yun-Yu Tseng, M Angeles Rabadán, Shefali Krishna, Michael Overholtzer, Gaudenz Danuser, Alan Hall
ABSTRACT
Efficient collective migration depends on a balance between contractility and cytoskeletal rearrangements, adhesion, and mechanical cell-cell communication, all controlled by GTPases of the RHO family. By comprehensive screening of guanine nucleotide exchange factors (GEFs) in human bronchial epithelial cell monolayers, we identified GEFs that are required for collective migration at large, such as SOS1 and β-PIX, and RHOA GEFs that are implicated in intercellular communication. Down-regulation of the latter GEFs differentially enhanced front-to-back propagation of guidance cues through the monolayer and was mirrored by down-regulation of RHOA expression and myosin II activity. Phenotype-based clustering of knockdown behaviors identified RHOA-ARHGEF18 and ARHGEF3-ARHGEF28-ARHGEF11 clusters, indicating that the latter may signal through other RHO-family GTPases. Indeed, knockdown of RHOC produced an intermediate between the two phenotypes. We conclude that for effective collective migration, the RHOA-GEFs → RHOA/C → actomyosin pathways must be optimally tuned to compromise between generation of motility forces and restriction of intercellular communication.
Sigma-Aldrich
MISSION® pLKO.1-puro Non-Mammalian shRNA Control Plasmid DNA, Targets no known mammalian genes
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting human TRIO
Pricing and availability is not currently available.