Merck
  • LY6/PLAUR domain containing 3 has a role in the maintenance of colorectal cancer stem-like cells.
LY6/PLAUR domain containing 3 has a role in the maintenance of colorectal cancer stem-like cells.
Biochemical and biophysical research communications (2017-02-28)
Liming Wang, Yoshihiko Hirohashi, Tadashi Ogawa, Min Shen, Rui Takeda, Aiko Murai, Eri Yamamoto, Terufumi Kubo, Munehide Nakatsugawa, Takayuki Kanaseki, Tomohide Tsukahara, Toshihiko Nishidate, Kenji Okita, Goro Kutomi, Noriyuki Sato, Ichiro Takemasa, Toshihiko Torigoe
ABSTRACT
Colorectal carcinoma (CRC) is one of the most frequently diagnosed cancers and the leading cause of cancer-related death for both men and women. Recent studies have revealed that a small sub-population of cancer cells, termed cancer stem-like cells (CSCs)/cancer-initiating cells (CICs), are endowed with tumor-initiating ability, self-renewal ability and differentiation ability. CSCs/CICs are resistant to current therapies including chemotherapy and radiotherapy. Thus, CSCs/CICs are responsible for recurrence and metastasis, and eradication of CSCs/CICs is essential to cure cancer. In this study, we isolated CR-CSCs/CICs as sphere-cultured cells and found that a product derived from LY6/PLAUR domain containing 3 (LYPD3) is preferentially expressed in CSCs/CICs. Gene overexpression and gene knockdown experiments revealed that LYPD3 has a role in the maintenance of CR-CSCs/CICs. The findings provide a novel molecular insight into CR-CSCs/CICs.
Sigma-Aldrich
MISSION® esiRNA, targeting human LYPD3