Merck
  • Microcystin-LR bioconcentration induces antioxidant responses in the digestive gland of two marine bivalves Crassostrea gigas and Mytilus edulis.
Microcystin-LR bioconcentration induces antioxidant responses in the digestive gland of two marine bivalves Crassostrea gigas and Mytilus edulis.
Aquatic toxicology (Amsterdam, Netherlands) (2017-05-14)
Young Dae Kim, Won Jin Kim, Yun Kyung Shin, Do-Hee Lee, Youn-Jung Kim, Jang Kyun Kim, Jae-Sung Rhee
ABSTRACT
Microcystins (MCs) are a major group of potent cyanobacterial toxins found in freshwater and even brackish waterbodies. To understand the putative correlation between bioconcentration of MCs and antioxidant responses of the digestive gland of bivalves, Pacific oyster Crassostrea gigas and blue mussel Mytilus edulis were exposed to different concentrations (0.1, 1, 10 and 20μgL
Sigma-Aldrich
SOD Assay Kit, sufficient for 500 tests
Sigma-Aldrich
Glutathione Peroxidase Cellular Activity Assay Kit, Sufficient for 100 colorimetric tests
Sigma-Aldrich
Glutathione Reductase Assay Kit, Sufficient for 100 colorimetric tests