Merck
CSFV proliferation is associated with GBF1 and Rab2.
Journal of biosciences (2017-02-24)
Wulong Liang, Minping Zheng, Changlei Bao, Yanming Zhang
ABSTRACT
The Golgi apparatus and its resident proteins are utilized and regulated by viruses to facilitate their proliferation. In this study, we investigated Classical swine fever virus (CSFV) proliferation when the function of the Golgi was disturbed. Golgi function was disturbed using chemical inhibitors, namely, brefeldin A (BFA) and golgicide A (GCA), and RNA interfering targets, such as the Golgi-specific BFA-resistance guanine nucleotide exchange factor 1 (GBF1) and Rab2 GTPases. CSFV proliferation was significantly inhibited during RNA replication and viral particle generation after BFA and GCA treatment. CSFV multiplication dynamics were retarded in cells transfected with GBF1 and Rab2 shRNA. Furthermore, CSFV proliferation was promoted by GBF1 and Rab2 overexpression using a lentiviral system. Hence, Golgi function is important for CSFV multiplication, and GBF1 and Rab2 participate in CSFV proliferation. Further studies must investigate Golgi-resident proteins to elucidate the mechanism underlying CSFV replication.
Sigma-Aldrich
Anti-Pig IgG (whole molecule)−FITC antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Golgicide A, ≥98% (HPLC)