Merck
  • Highly Efficient Luminescent Metal-Organic Framework for the Simultaneous Detection and Removal of Heavy Metals from Water.
Highly Efficient Luminescent Metal-Organic Framework for the Simultaneous Detection and Removal of Heavy Metals from Water.
ACS applied materials & interfaces (2016-10-14)
Nathan D Rudd, Hao Wang, Erika M A Fuentes-Fernandez, Simon J Teat, Feng Chen, Gene Hall, Yves J Chabal, Jing Li
ABSTRACT
We have designed and synthesized an isoreticular series of luminescent metal-organic frameworks (LMOFs) by incorporating a strongly emissive molecular fluorophore and functionally diverse colinkers into Zn-based structures. The three-dimensional porous networks of LMOF-261, -262, and -263 represent a unique/new type of nets, classified as a 2-nodal, (4,4)-c net (mot-e type) with 4-fold, class IIIa interpenetration. All compounds crystallize in a body-centered tetragonal crystal system (space group I4
Sigma-Aldrich
1,1,2,2-Tetrakis(4-bromophenyl)ethylene