Merck
  • Pharmacological Characterization of a Novel Beta 3 Adrenergic Agonist, Vibegron: Evaluation of Antimuscarinic Receptor Selectivity for Combination Therapy for Overactive Bladder.
Pharmacological Characterization of a Novel Beta 3 Adrenergic Agonist, Vibegron: Evaluation of Antimuscarinic Receptor Selectivity for Combination Therapy for Overactive Bladder.
The Journal of pharmacology and experimental therapeutics (2016-12-15)
J Di Salvo, H Nagabukuro, L A Wickham, C Abbadie, J A DeMartino, A Fitzmaurice, L Gichuru, A Kulick, M J Donnelly, N Jochnowitz, A L Hurley, A Pereira, A Sanfiz, G Veronin, K Villa, J Woods, B Zamlynny, E Zycband, G M Salituro, T Frenkl, A E Weber, S D Edmondson, M Struthers
ABSTRACT
Although the physiologic role of muscarinic receptors in bladder function and the therapeutic efficacy of muscarinic antagonists for the treatment of overactive bladder are well established, the role of β
Sigma-Aldrich
Darifenacin hydrobromide, ≥98% (HPLC)
Sigma-Aldrich
Methoctramine hydrate, ≥97% (NMR), solid
Sigma-Aldrich
Free Glycerol Determination Kit, 1 kit sufficient for 1000 reactions
Sigma-Aldrich
CL 316,243 hydrate, ≥98% (HPLC), powder