MilliporeSigma
  • Neutrophils are dispensable in the modulation of T cell immunity against cutaneous HSV-1 infection.
Neutrophils are dispensable in the modulation of T cell immunity against cutaneous HSV-1 infection.
Scientific reports (2017-01-24)
Jyh Liang Hor, William R Heath, Scott N Mueller
ABSTRACT
Neutrophils rapidly infiltrate sites of inflammation during peripheral infection or tissue injury. In addition to their well described roles as pro-inflammatory phagocytes responsible for pathogen clearance, recent studies have demonstrated a broader functional repertoire including mediating crosstalk between innate and adaptive arms of the immune system. Specifically, neutrophils have been proposed to mediate antigen transport to lymph nodes (LN) to modulate T cell priming and to influence T cell migration to infected tissues. Using a mouse model of cutaneous herpes simplex virus type 1 (HSV-1) infection we explored potential contributions of neutrophils toward anti-viral immunity. While a transient, early influx of neutrophils was triggered by dermal scarification, we did not detect migration of neutrophils from the skin to LN. Furthermore, despite recruitment of neutrophils into LN from the blood, priming and expansion of CD4